Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63939169-63939442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK2
Entrez Gene ID
23604
Gene Name
death associated protein kinase 2
Gene Aliases
DRP-1, DRP1
UniGene
Hs.237886
Gene Chromosome Location
Chr.15: 63907036-64046470 on Build GRCh38
Transcripts
NM_014326.3 XM_011521413.2 XM_011521414.1 XM_011521415.2 XM_011521416.2 XM_011521417.1 XM_011521421.2 XM_017022043.1 XM_017022044.1 XM_017022045.1 XM_017022046.1 XM_017022047.1 XM_017022048.1 XM_017022049.1 XM_017022050.1 XM_017022051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3887291 c.497A>C p.H166P Chr.15: 63939318-63939318 on Build GRCh38
274735 c.570G>A p.T190T Chr.15: 63939245-63939245 on Build GRCh38
5804614 c.512A>T p.D171V Chr.15: 63939303-63939303 on Build GRCh38
5342610 c.532A>C p.I178L Chr.15: 63939283-63939283 on Build GRCh38
99056 c.528C>G p.H176Q Chr.15: 63939287-63939287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746749952 Chr.15: 63939290-63939290 on Build GRCh38
rs781019991 Chr.15: 63939404-63939404 on Build GRCh38
rs75770249 Chr.15: 63939379-63939379 on Build GRCh38
rs780185443 Chr.15: 63939384-63939384 on Build GRCh38
rs778067790 Chr.15: 63939192-63939192 on Build GRCh38
rs201994664 Chr.15: 63939340-63939340 on Build GRCh38
rs1036578397 Chr.15: 63939397-63939397 on Build GRCh38
rs772751720 Chr.15: 63939322-63939322 on Build GRCh38
rs563315259 Chr.15: 63939317-63939317 on Build GRCh38
rs200347768 Chr.15: 63939406-63939406 on Build GRCh38
rs75862994 Chr.15: 63939380-63939380 on Build GRCh38
rs749266624 Chr.15: 63939199-63939199 on Build GRCh38
rs368893784 Chr.15: 63939206-63939206 on Build GRCh38
rs760670118 Chr.15: 63939228-63939228 on Build GRCh38
rs761163560 Chr.15: 63939234-63939234 on Build GRCh38
rs768332970 Chr.15: 63939380-63939380 on Build GRCh38
rs370730920 Chr.15: 63939367-63939367 on Build GRCh38
rs1018727274 Chr.15: 63939258-63939258 on Build GRCh38
rs995496297 Chr.15: 63939198-63939198 on Build GRCh38
rs752455081 Chr.15: 63939242-63939242 on Build GRCh38
rs771903012 Chr.15: 63939219-63939219 on Build GRCh38
rs186891659 Chr.15: 63939287-63939287 on Build GRCh38
rs776364557 Chr.15: 63939380-63939381 on Build GRCh38
rs771541762 Chr.15: 63939348-63939350 on Build GRCh38
rs1049037874 Chr.15: 63939360-63939360 on Build GRCh38
rs776392199 Chr.15: 63939292-63939292 on Build GRCh38
rs750486428 Chr.15: 63939381-63939381 on Build GRCh38
rs551687423 Chr.15: 63939306-63939306 on Build GRCh38
rs745694509 Chr.15: 63939211-63939211 on Build GRCh38
rs767646127 Chr.15: 63939405-63939406 on Build GRCh38
rs756699976 Chr.15: 63939256-63939256 on Build GRCh38
rs149502968 Chr.15: 63939246-63939246 on Build GRCh38
rs567209501 Chr.15: 63939406-63939406 on Build GRCh38
rs752826388 Chr.15: 63939406-63939408 on Build GRCh38
rs764973476 Chr.15: 63939235-63939235 on Build GRCh38
rs779027761 Chr.15: 63939277-63939277 on Build GRCh38
rs756661231 Chr.15: 63939190-63939190 on Build GRCh38
rs889525456 Chr.15: 63939418-63939418 on Build GRCh38
rs539034378 Chr.15: 63939281-63939281 on Build GRCh38
rs757751054 Chr.15: 63939376-63939376 on Build GRCh38
rs749996059 Chr.15: 63939241-63939241 on Build GRCh38
rs966172470 Chr.15: 63939273-63939273 on Build GRCh38
rs377220629 Chr.15: 63939364-63939364 on Build GRCh38
rs868275663 Chr.15: 63939295-63939295 on Build GRCh38
rs768038957 Chr.15: 63939243-63939243 on Build GRCh38
rs768392118 Chr.15: 63939400-63939400 on Build GRCh38
rs753979326 Chr.15: 63939269-63939269 on Build GRCh38
rs764679085 Chr.15: 63939370-63939370 on Build GRCh38
rs73452775 Chr.15: 63939417-63939417 on Build GRCh38
rs766120131 Chr.15: 63939350-63939350 on Build GRCh38
rs774996778 Chr.15: 63939377-63939378 on Build GRCh38
rs771186341 Chr.15: 63939201-63939201 on Build GRCh38
rs757231979 Chr.15: 63939270-63939270 on Build GRCh38
rs778355750 Chr.15: 63939259-63939259 on Build GRCh38
rs967994152 Chr.15: 63939351-63939351 on Build GRCh38
rs372823645 Chr.15: 63939335-63939335 on Build GRCh38
rs758631998 Chr.15: 63939382-63939382 on Build GRCh38
rs775234046 Chr.15: 63939220-63939220 on Build GRCh38
rs940912517 Chr.15: 63939392-63939392 on Build GRCh38
rs750392202 Chr.15: 63939397-63939398 on Build GRCh38
rs768587064 Chr.15: 63939291-63939291 on Build GRCh38
rs768566447 Chr.15: 63939231-63939231 on Build GRCh38
rs777665315 Chr.15: 63939286-63939286 on Build GRCh38
rs765185832 Chr.15: 63939389-63939390 on Build GRCh38
rs776220153 Chr.15: 63939232-63939232 on Build GRCh38
rs760256945 Chr.15: 63939379-63939380 on Build GRCh38
rs778926902 Chr.15: 63939377-63939377 on Build GRCh38
rs745380240 Chr.15: 63939223-63939233 on Build GRCh38
rs549068017 Chr.15: 63939405-63939405 on Build GRCh38
rs772658512 Chr.15: 63939412-63939412 on Build GRCh38
rs746839808 Chr.15: 63939393-63939393 on Build GRCh38

View Full Product Details