Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45120808-45121061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP3
Entrez Gene ID
2287
Gene Name
FK506 binding protein 3
Gene Aliases
FKBP-25, FKBP-3, FKBP25, PPIase
UniGene
Hs.509226
Gene Chromosome Location
Chr.14: 45115599-45134529 on Build GRCh38
Transcripts
NM_002013.3 XM_011536565.2 XM_017021092.1 XM_017021093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664257 c.512T>C p.V171A Chr.14: 45120897-45120897 on Build GRCh38
6326794 c.502G>A p.V168I Chr.14: 45120907-45120907 on Build GRCh38
6326132 c.495G>A p.K165K Chr.14: 45120914-45120914 on Build GRCh38
6251983 c.481C>A p.P161T Chr.14: 45120928-45120928 on Build GRCh38
7223959 c.462G>C p.K154N Chr.14: 45120947-45120947 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556866469 Chr.14: 45120885-45120885 on Build GRCh38
rs760289685 Chr.14: 45120921-45120921 on Build GRCh38
rs772262482 Chr.14: 45120997-45120999 on Build GRCh38
rs779246024 Chr.14: 45120939-45120941 on Build GRCh38
rs746273839 Chr.14: 45120995-45120998 on Build GRCh38
rs778000901 Chr.14: 45120877-45120877 on Build GRCh38
rs745902703 Chr.14: 45121010-45121010 on Build GRCh38
rs758412192 Chr.14: 45120951-45120951 on Build GRCh38
rs779967895 Chr.14: 45120968-45120968 on Build GRCh38
rs758130720 Chr.14: 45120867-45120867 on Build GRCh38
rs903507160 Chr.14: 45120852-45120852 on Build GRCh38
rs1019071958 Chr.14: 45120982-45120982 on Build GRCh38
rs759907896 Chr.14: 45120904-45120904 on Build GRCh38
rs561308436 Chr.14: 45120898-45120898 on Build GRCh38
rs771413110 Chr.14: 45120894-45120894 on Build GRCh38
rs866682962 Chr.14: 45120974-45120974 on Build GRCh38
rs751467067 Chr.14: 45120988-45120988 on Build GRCh38
rs377144510 Chr.14: 45120869-45120869 on Build GRCh38
rs757456740 Chr.14: 45120855-45120855 on Build GRCh38
rs765152978 Chr.14: 45120941-45120941 on Build GRCh38
rs1007648274 Chr.14: 45120908-45120908 on Build GRCh38
rs369567635 Chr.14: 45120895-45120895 on Build GRCh38
rs750351667 Chr.14: 45120945-45120945 on Build GRCh38
rs568810956 Chr.14: 45120913-45120913 on Build GRCh38
rs772446215 Chr.14: 45120911-45120911 on Build GRCh38
rs761239388 Chr.14: 45120937-45120937 on Build GRCh38
rs749386203 Chr.14: 45120889-45120889 on Build GRCh38
rs144723350 Chr.14: 45120868-45120868 on Build GRCh38
rs781297946 Chr.14: 45120897-45120897 on Build GRCh38
rs757210950 Chr.14: 45120845-45120845 on Build GRCh38
rs753329047 Chr.14: 45120935-45120935 on Build GRCh38
rs747168359 Chr.14: 45120835-45120835 on Build GRCh38
rs966650171 Chr.14: 45120992-45120993 on Build GRCh38
rs763803360 Chr.14: 45120933-45120933 on Build GRCh38
rs745615147 Chr.14: 45120862-45120862 on Build GRCh38

View Full Product Details