Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13356618-13356891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
UniGene
Hs.65734
Gene Chromosome Location
Chr.11: 13276613-13387268 on Build GRCh38
Transcripts
NM_001030272.2 NM_001030273.2 NM_001178.5 NM_001297719.1 NM_001297722.1 NM_001297724.1 XM_006718233.3 XM_006718234.1 XM_011520105.2 XM_011520107.2 XM_011520108.1 XM_011520109.1 XM_011520110.1 XM_011520111.1 XM_011520112.2 XM_011520113.1 XM_017017738.1 XM_017017739.1 XM_017017740.1 XM_017017741.1 XM_017017742.1 XM_017017743.1 XM_017017744.1 XM_017017745.1 XM_017017746.1 XM_017017747.1 XM_017017748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3676117 c.179+6A>G p.? Chr.11: 13356783-13356783 on Build GRCh38
3676116 c.179+6A>G p.? Chr.11: 13356783-13356783 on Build GRCh38
925070 c.163G>T p.D55Y Chr.11: 13356761-13356761 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780425624 Chr.11: 13356842-13356842 on Build GRCh38
rs74577097 Chr.11: 13356676-13356676 on Build GRCh38
rs776656663 Chr.11: 13356724-13356724 on Build GRCh38
rs778487473 Chr.11: 13356691-13356691 on Build GRCh38
rs746238303 Chr.11: 13356693-13356693 on Build GRCh38
rs761943304 Chr.11: 13356744-13356744 on Build GRCh38
rs376385769 Chr.11: 13356826-13356826 on Build GRCh38
rs61882386 Chr.11: 13356825-13356825 on Build GRCh38
rs908179641 Chr.11: 13356701-13356701 on Build GRCh38
rs753385613 Chr.11: 13356823-13356823 on Build GRCh38
rs747411474 Chr.11: 13356848-13356848 on Build GRCh38
rs140130779 Chr.11: 13356772-13356772 on Build GRCh38
rs892785524 Chr.11: 13356686-13356686 on Build GRCh38
rs372870035 Chr.11: 13356859-13356859 on Build GRCh38
rs996154055 Chr.11: 13356845-13356845 on Build GRCh38
rs764176518 Chr.11: 13356820-13356820 on Build GRCh38
rs772326388 Chr.11: 13356704-13356704 on Build GRCh38
rs776005531 Chr.11: 13356708-13356708 on Build GRCh38
rs767240427 Chr.11: 13356806-13356806 on Build GRCh38
rs752299267 Chr.11: 13356727-13356729 on Build GRCh38
rs748196475 Chr.11: 13356867-13356867 on Build GRCh38
rs747532981 Chr.11: 13356714-13356714 on Build GRCh38
rs745518415 Chr.11: 13356829-13356829 on Build GRCh38
rs368980029 Chr.11: 13356718-13356718 on Build GRCh38
rs374861187 Chr.11: 13356719-13356719 on Build GRCh38
rs932586600 Chr.11: 13356817-13356817 on Build GRCh38
rs368051377 Chr.11: 13356811-13356811 on Build GRCh38
rs758737644 Chr.11: 13356839-13356839 on Build GRCh38
rs749730533 Chr.11: 13356725-13356726 on Build GRCh38
rs147480315 Chr.11: 13356677-13356677 on Build GRCh38
rs769780672 Chr.11: 13356868-13356868 on Build GRCh38
rs1003060808 Chr.11: 13356775-13356775 on Build GRCh38
rs765496273 Chr.11: 13356767-13356767 on Build GRCh38
rs769220676 Chr.11: 13356770-13356770 on Build GRCh38
rs769126334 Chr.11: 13356856-13356856 on Build GRCh38
rs939699729 Chr.11: 13356703-13356703 on Build GRCh38
rs371906248 Chr.11: 13356815-13356815 on Build GRCh38
rs759319968 Chr.11: 13356805-13356805 on Build GRCh38
rs1009866609 Chr.11: 13356695-13356695 on Build GRCh38

View Full Product Details