Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49239156-49239407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
UniGene
Hs.122006
Gene Chromosome Location
Chr.15: 49155757-49367883 on Build GRCh38
Transcripts
NM_001001556.2 NM_001289030.1 NM_001289031.1 NM_002044.3 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720461.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022062.1 XM_017022063.1 XM_017022064.1 XM_017022065.1 XM_017022066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344423 c.400G>T p.V134L Chr.15: 49239263-49239263 on Build GRCh38
4754828 c.473C>T p.T158M Chr.15: 49239336-49239336 on Build GRCh38
6554955 c.384A>T p.P128P Chr.15: 49239280-49239280 on Build GRCh38
3501876 c.467C>T p.S156F Chr.15: 49239363-49239363 on Build GRCh38
6249078 c.385G>T p.G129* Chr.15: 49239248-49239248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766251479 Chr.15: 49239345-49239345 on Build GRCh38
rs1032901305 Chr.15: 49239303-49239303 on Build GRCh38
rs374871677 Chr.15: 49239329-49239329 on Build GRCh38
rs779555164 Chr.15: 49239285-49239285 on Build GRCh38
rs141479055 Chr.15: 49239340-49239340 on Build GRCh38
rs764352990 Chr.15: 49239364-49239364 on Build GRCh38
rs967098843 Chr.15: 49239338-49239338 on Build GRCh38
rs768327888 Chr.15: 49239321-49239321 on Build GRCh38
rs573578687 Chr.15: 49239313-49239313 on Build GRCh38
rs756310089 Chr.15: 49239197-49239197 on Build GRCh38
rs148234101 Chr.15: 49239230-49239230 on Build GRCh38
rs531037984 Chr.15: 49239295-49239295 on Build GRCh38
rs201859172 Chr.15: 49239374-49239374 on Build GRCh38
rs747946698 Chr.15: 49239335-49239335 on Build GRCh38
rs201904223 Chr.15: 49239288-49239288 on Build GRCh38
rs983703786 Chr.15: 49239259-49239259 on Build GRCh38
rs372905837 Chr.15: 49239241-49239241 on Build GRCh38
rs757666021 Chr.15: 49239210-49239210 on Build GRCh38
rs764253332 Chr.15: 49239201-49239201 on Build GRCh38
rs758830352 Chr.15: 49239265-49239265 on Build GRCh38
rs544545223 Chr.15: 49239337-49239337 on Build GRCh38
rs199837168 Chr.15: 49239209-49239209 on Build GRCh38
rs769779855 Chr.15: 49239336-49239336 on Build GRCh38
rs760709272 Chr.15: 49239363-49239363 on Build GRCh38
rs947815363 Chr.15: 49239351-49239351 on Build GRCh38
rs775598902 Chr.15: 49239356-49239356 on Build GRCh38
rs1030320436 Chr.15: 49239200-49239200 on Build GRCh38

View Full Product Details