Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16595109-16595380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZL
Entrez Gene ID
1618
Gene Name
deleted in azoospermia like
Gene Aliases
DAZH, DAZL1, DAZLA, SPGYLA
UniGene
Hs.131179
Gene Chromosome Location
Chr.3: 16586792-16605499 on Build GRCh38
Transcripts
NM_001190811.1 NM_001351.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6263442 c.570+10A>G p.? Chr.3: 16595304-16595304 on Build GRCh38
6264445 c.570+6T>G p.? Chr.3: 16595308-16595308 on Build GRCh38
4676246 c.513C>T p.Y171Y Chr.3: 16595371-16595371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746737596 Chr.3: 16595323-16595323 on Build GRCh38
rs748885412 Chr.3: 16595354-16595354 on Build GRCh38
rs757402516 Chr.3: 16595300-16595300 on Build GRCh38
rs748066712 Chr.3: 16595332-16595332 on Build GRCh38
rs754777205 Chr.3: 16595324-16595324 on Build GRCh38
rs569380170 Chr.3: 16595295-16595295 on Build GRCh38
rs939063519 Chr.3: 16595169-16595169 on Build GRCh38
rs369007495 Chr.3: 16595336-16595336 on Build GRCh38
rs766102896 Chr.3: 16595279-16595279 on Build GRCh38
rs774845129 Chr.3: 16595282-16595282 on Build GRCh38
rs764116038 Chr.3: 16595299-16595299 on Build GRCh38
rs758429614 Chr.3: 16595281-16595281 on Build GRCh38
rs2254783 Chr.3: 16595166-16595166 on Build GRCh38
rs780140951 Chr.3: 16595290-16595290 on Build GRCh38
rs756173358 Chr.3: 16595298-16595298 on Build GRCh38
rs1054808096 Chr.3: 16595269-16595269 on Build GRCh38
rs768071691 Chr.3: 16595286-16595286 on Build GRCh38
rs763567586 Chr.3: 16595186-16595186 on Build GRCh38
rs374944997 Chr.3: 16595308-16595308 on Build GRCh38
rs906225397 Chr.3: 16595168-16595168 on Build GRCh38
rs772572791 Chr.3: 16595352-16595352 on Build GRCh38
rs2303590 Chr.3: 16595278-16595278 on Build GRCh38
rs368616887 Chr.3: 16595267-16595267 on Build GRCh38
rs760045685 Chr.3: 16595283-16595283 on Build GRCh38

View Full Product Details