Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123241295-123241520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf76
Entrez Gene ID
84933
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123219956-123241398 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Gene Symbol
ZHX1-C8orf76
Entrez Gene ID
100533106
Gene Name
ZHX1-C8orf76 readthrough
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123226189-123274487 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
749493 c.52G>C p.E18Q Chr.8: 123241295-123241295 on Build GRCh38
253692 c.37G>A p.E13K Chr.8: 123241310-123241310 on Build GRCh38
749492 c.40G>C p.D14H Chr.8: 123241307-123241307 on Build GRCh38
239096 c.24C>G p.F8L Chr.8: 123241323-123241323 on Build GRCh38
5016788 c.14G>C p.C5S Chr.8: 123241333-123241333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759602007 Chr.8: 123241330-123241330 on Build GRCh38
rs745703663 Chr.8: 123241393-123241393 on Build GRCh38
rs764537892 Chr.8: 123241360-123241360 on Build GRCh38
rs748074864 Chr.8: 123241365-123241366 on Build GRCh38
rs760000812 Chr.8: 123241365-123241365 on Build GRCh38
rs757074804 Chr.8: 123241342-123241342 on Build GRCh38
rs770059188 Chr.8: 123241358-123241358 on Build GRCh38
rs778870550 Chr.8: 123241356-123241358 on Build GRCh38
rs981304421 Chr.8: 123241460-123241460 on Build GRCh38
rs982159018 Chr.8: 123241401-123241401 on Build GRCh38
rs576491864 Chr.8: 123241338-123241338 on Build GRCh38
rs777599549 Chr.8: 123241351-123241351 on Build GRCh38
rs764625938 Chr.8: 123241335-123241335 on Build GRCh38
rs754651314 Chr.8: 123241369-123241369 on Build GRCh38
rs35017855 Chr.8: 123241342-123241343 on Build GRCh38
rs747705071 Chr.8: 123241344-123241344 on Build GRCh38
rs771018919 Chr.8: 123241378-123241379 on Build GRCh38
rs888783938 Chr.8: 123241485-123241485 on Build GRCh38
rs767082519 Chr.8: 123241359-123241359 on Build GRCh38
rs1027967532 Chr.8: 123241364-123241364 on Build GRCh38
rs774736327 Chr.8: 123241361-123241361 on Build GRCh38
rs187553021 Chr.8: 123241449-123241449 on Build GRCh38
rs771141626 Chr.8: 123241325-123241325 on Build GRCh38
rs1050670116 Chr.8: 123241479-123241479 on Build GRCh38
rs78443181 Chr.8: 123241372-123241372 on Build GRCh38
rs777706231 Chr.8: 123241373-123241373 on Build GRCh38
rs757284767 Chr.8: 123241381-123241381 on Build GRCh38
rs575551607 Chr.8: 123241454-123241454 on Build GRCh38
rs751913898 Chr.8: 123241337-123241337 on Build GRCh38
rs1020217353 Chr.8: 123241407-123241407 on Build GRCh38
rs991377827 Chr.8: 123241366-123241366 on Build GRCh38
rs374375604 Chr.8: 123241356-123241356 on Build GRCh38
rs767772069 Chr.8: 123241363-123241363 on Build GRCh38
rs772100489 Chr.8: 123241346-123241346 on Build GRCh38
rs760090989 Chr.8: 123241477-123241477 on Build GRCh38
rs542152915 Chr.8: 123241436-123241436 on Build GRCh38
rs776738933 Chr.8: 123241328-123241328 on Build GRCh38
rs1053106977 Chr.8: 123241322-123241322 on Build GRCh38
rs1052639575 Chr.8: 123241455-123241455 on Build GRCh38
rs765305157 Chr.8: 123241332-123241332 on Build GRCh38
rs746767634 Chr.8: 123241352-123241352 on Build GRCh38
rs938073647 Chr.8: 123241487-123241487 on Build GRCh38
rs1010557473 Chr.8: 123241334-123241334 on Build GRCh38
rs893982943 Chr.8: 123241466-123241466 on Build GRCh38
rs754747617 Chr.8: 123241343-123241343 on Build GRCh38
rs753440010 Chr.8: 123241339-123241339 on Build GRCh38
rs1018747176 Chr.8: 123241349-123241349 on Build GRCh38
rs780835654 Chr.8: 123241388-123241388 on Build GRCh38
rs889536526 Chr.8: 123241360-123241361 on Build GRCh38
rs769964611 Chr.8: 123241398-123241398 on Build GRCh38
rs368425076 Chr.8: 123241370-123241370 on Build GRCh38
rs746923996 Chr.8: 123241380-123241380 on Build GRCh38
rs752840233 Chr.8: 123241333-123241333 on Build GRCh38
rs772190463 Chr.8: 123241320-123241320 on Build GRCh38
rs773115630 Chr.8: 123241323-123241323 on Build GRCh38
rs927994112 Chr.8: 123241481-123241481 on Build GRCh38
rs774902643 Chr.8: 123241362-123241362 on Build GRCh38

View Full Product Details