Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135545218-135545491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS6L
Entrez Gene ID
203522
Gene Name
integrator complex subunit 6 like
Gene Aliases
DDX26B
UniGene
Hs.496829
Gene Chromosome Location
Chr.X: 135520630-135582535 on Build GRCh38
Transcripts
NM_182540.4 XM_005262394.2 XM_006724740.2 XM_006724741.3 XM_011531311.2 XM_017029342.1 XM_017029343.1 XM_017029344.1 XM_017029345.1 XM_017029346.1 XM_017029347.1 XM_017029348.1 XM_017029349.1

SNP Detail

SNP ID Chromosome Location
rs781924630 Chr.X: 135545335-135545335 on Build GRCh38
rs782069660 Chr.X: 135545346-135545346 on Build GRCh38
rs1020464180 Chr.X: 135545258-135545258 on Build GRCh38
rs782418168 Chr.X: 135545384-135545384 on Build GRCh38
rs782311581 Chr.X: 135545455-135545455 on Build GRCh38
rs782069427 Chr.X: 135545459-135545459 on Build GRCh38
rs376094163 Chr.X: 135545270-135545270 on Build GRCh38
rs782117890 Chr.X: 135545452-135545452 on Build GRCh38
rs1009952772 Chr.X: 135545245-135545245 on Build GRCh38
rs781880714 Chr.X: 135545415-135545415 on Build GRCh38
rs374719028 Chr.X: 135545461-135545461 on Build GRCh38
rs782298075 Chr.X: 135545306-135545306 on Build GRCh38
rs782015330 Chr.X: 135545447-135545447 on Build GRCh38
rs781956217 Chr.X: 135545458-135545458 on Build GRCh38

View Full Product Details