Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1331644-1331915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
UniGene
Hs.444673
Gene Chromosome Location
Chr.5: 1317744-1345070 on Build GRCh38
Transcripts
NM_030782.4 XM_011514144.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32738 c.939G>T p.K313N Chr.5: 1331836-1331836 on Build GRCh38
7179982 c.945G>C p.K315N Chr.5: 1331830-1331830 on Build GRCh38
122203 c.957C>G p.I319M Chr.5: 1331818-1331818 on Build GRCh38
4443000 c.928A>G p.S310G Chr.5: 1331847-1331847 on Build GRCh38
3850116 c.944A>G p.K315R Chr.5: 1331831-1331831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776960207 Chr.5: 1331772-1331772 on Build GRCh38
rs752485426 Chr.5: 1331793-1331793 on Build GRCh38
rs777316786 Chr.5: 1331750-1331750 on Build GRCh38
rs917428835 Chr.5: 1331709-1331709 on Build GRCh38
rs755244914 Chr.5: 1331841-1331841 on Build GRCh38
rs375454190 Chr.5: 1331839-1331839 on Build GRCh38
rs777642149 Chr.5: 1331807-1331807 on Build GRCh38
rs778851318 Chr.5: 1331751-1331751 on Build GRCh38
rs373925542 Chr.5: 1331792-1331792 on Build GRCh38
rs759783318 Chr.5: 1331893-1331893 on Build GRCh38
rs111874283 Chr.5: 1331775-1331775 on Build GRCh38
rs199975249 Chr.5: 1331851-1331851 on Build GRCh38
rs756050018 Chr.5: 1331803-1331803 on Build GRCh38
rs940099232 Chr.5: 1331746-1331746 on Build GRCh38
rs35184924 Chr.5: 1331696-1331697 on Build GRCh38
rs771003320 Chr.5: 1331863-1331863 on Build GRCh38
rs553002482 Chr.5: 1331864-1331864 on Build GRCh38
rs759288082 Chr.5: 1331791-1331791 on Build GRCh38
rs142281891 Chr.5: 1331818-1331818 on Build GRCh38
rs773309700 Chr.5: 1331780-1331780 on Build GRCh38
rs143530499 Chr.5: 1331826-1331828 on Build GRCh38
rs372893688 Chr.5: 1331783-1331783 on Build GRCh38
rs769065743 Chr.5: 1331768-1331768 on Build GRCh38
rs371510655 Chr.5: 1331787-1331787 on Build GRCh38
rs926250775 Chr.5: 1331742-1331742 on Build GRCh38
rs747323472 Chr.5: 1331766-1331766 on Build GRCh38
rs974816150 Chr.5: 1331666-1331666 on Build GRCh38
rs773765324 Chr.5: 1331852-1331852 on Build GRCh38
rs149130584 Chr.5: 1331764-1331764 on Build GRCh38
rs2306194 Chr.5: 1331719-1331719 on Build GRCh38
rs538224811 Chr.5: 1331693-1331693 on Build GRCh38
rs753387938 Chr.5: 1331816-1331816 on Build GRCh38
rs781595771 Chr.5: 1331842-1331842 on Build GRCh38
rs751793114 Chr.5: 1331788-1331788 on Build GRCh38
rs950868306 Chr.5: 1331663-1331663 on Build GRCh38
rs988051768 Chr.5: 1331797-1331797 on Build GRCh38
rs555554731 Chr.5: 1331850-1331850 on Build GRCh38
rs571406390 Chr.5: 1331736-1331736 on Build GRCh38
rs1002173904 Chr.5: 1331761-1331761 on Build GRCh38
rs778645980 Chr.5: 1331820-1331820 on Build GRCh38
rs774508335 Chr.5: 1331869-1331869 on Build GRCh38
rs886296301 Chr.5: 1331737-1331737 on Build GRCh38
rs770706553 Chr.5: 1331754-1331754 on Build GRCh38
rs749307820 Chr.5: 1331860-1331860 on Build GRCh38

View Full Product Details