Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205058403-205058648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN2
Entrez Gene ID
6900
Gene Name
contactin 2
Gene Aliases
AXT, FAME5, TAG-1, TAX, TAX1
UniGene
Hs.519220
Gene Chromosome Location
Chr.1: 205043160-205078043 on Build GRCh38
Transcripts
NM_005076.3 XM_017002198.1 XM_017002199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733816 c.415G>A p.E139K Chr.1: 205058591-205058591 on Build GRCh38
4606701 c.401A>T p.E134V Chr.1: 205058577-205058577 on Build GRCh38
3746743 c.433G>A p.A145T Chr.1: 205058609-205058609 on Build GRCh38
2214839 c.445T>C p.W149R Chr.1: 205058621-205058621 on Build GRCh38
6343236 c.427G>T p.V143L Chr.1: 205058603-205058603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984041853 Chr.1: 205058548-205058548 on Build GRCh38
rs770526190 Chr.1: 205058461-205058461 on Build GRCh38
rs774281069 Chr.1: 205058599-205058599 on Build GRCh38
rs9787409 Chr.1: 205058433-205058433 on Build GRCh38
rs769264642 Chr.1: 205058474-205058474 on Build GRCh38
rs778209101 Chr.1: 205058458-205058458 on Build GRCh38
rs768539409 Chr.1: 205058497-205058497 on Build GRCh38
rs566378735 Chr.1: 205058451-205058451 on Build GRCh38
rs765324113 Chr.1: 205058523-205058523 on Build GRCh38
rs761641756 Chr.1: 205058508-205058508 on Build GRCh38
rs766545038 Chr.1: 205058510-205058510 on Build GRCh38
rs762796697 Chr.1: 205058496-205058496 on Build GRCh38
rs765748170 Chr.1: 205058530-205058532 on Build GRCh38
rs780555759 Chr.1: 205058466-205058466 on Build GRCh38
rs745602659 Chr.1: 205058471-205058471 on Build GRCh38
rs536047902 Chr.1: 205058448-205058448 on Build GRCh38
rs1008923321 Chr.1: 205058525-205058525 on Build GRCh38
rs759814182 Chr.1: 205058513-205058513 on Build GRCh38
rs768629152 Chr.1: 205058595-205058595 on Build GRCh38
rs567031902 Chr.1: 205058442-205058442 on Build GRCh38
rs780758694 Chr.1: 205058561-205058561 on Build GRCh38
rs776790948 Chr.1: 205058628-205058628 on Build GRCh38
rs758805948 Chr.1: 205058533-205058533 on Build GRCh38
rs745312005 Chr.1: 205058564-205058564 on Build GRCh38
rs779892361 Chr.1: 205058594-205058594 on Build GRCh38
rs752120567 Chr.1: 205058545-205058545 on Build GRCh38
rs199903175 Chr.1: 205058535-205058535 on Build GRCh38
rs868191357 Chr.1: 205058576-205058576 on Build GRCh38
rs755814341 Chr.1: 205058591-205058591 on Build GRCh38
rs2275697 Chr.1: 205058609-205058609 on Build GRCh38
rs966267940 Chr.1: 205058588-205058588 on Build GRCh38
rs543467450 Chr.1: 205058460-205058460 on Build GRCh38
rs148381903 Chr.1: 205058554-205058554 on Build GRCh38
rs143259493 Chr.1: 205058600-205058600 on Build GRCh38
rs753791279 Chr.1: 205058511-205058511 on Build GRCh38

View Full Product Details