Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137050315-137050554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866980 c.829G>T p.G277W Chr.9: 137050484-137050484 on Build GRCh38
4994054 c.829G>A p.G277R Chr.9: 137050484-137050484 on Build GRCh38
5359186 c.948G>A p.G316G Chr.9: 137050365-137050365 on Build GRCh38
4562573 c.932G>A p.R311Q Chr.9: 137050381-137050381 on Build GRCh38
3926476 c.807C>T p.G269G Chr.9: 137050506-137050506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370555871 Chr.9: 137050513-137050513 on Build GRCh38
rs774971518 Chr.9: 137050410-137050410 on Build GRCh38
rs748596985 Chr.9: 137050381-137050381 on Build GRCh38
rs1037086545 Chr.9: 137050477-137050477 on Build GRCh38
rs747460688 Chr.9: 137050496-137050498 on Build GRCh38
rs748902935 Chr.9: 137050475-137050475 on Build GRCh38
rs778135985 Chr.9: 137050463-137050468 on Build GRCh38
rs774134020 Chr.9: 137050426-137050426 on Build GRCh38
rs866672782 Chr.9: 137050516-137050516 on Build GRCh38
rs753979087 Chr.9: 137050441-137050441 on Build GRCh38
rs138882764 Chr.9: 137050342-137050342 on Build GRCh38
rs371567979 Chr.9: 137050512-137050512 on Build GRCh38
rs956231651 Chr.9: 137050347-137050347 on Build GRCh38
rs142156660 Chr.9: 137050448-137050448 on Build GRCh38
rs775013850 Chr.9: 137050474-137050474 on Build GRCh38
rs758600277 Chr.9: 137050455-137050455 on Build GRCh38
rs745407606 Chr.9: 137050470-137050470 on Build GRCh38
rs745356405 Chr.9: 137050402-137050402 on Build GRCh38
rs148619460 Chr.9: 137050533-137050533 on Build GRCh38
rs757134250 Chr.9: 137050357-137050357 on Build GRCh38
rs761747255 Chr.9: 137050427-137050427 on Build GRCh38
rs752765897 Chr.9: 137050515-137050515 on Build GRCh38
rs761587692 Chr.9: 137050496-137050496 on Build GRCh38
rs764129953 Chr.9: 137050343-137050343 on Build GRCh38
rs762624119 Chr.9: 137050411-137050411 on Build GRCh38
rs753851953 Chr.9: 137050365-137050365 on Build GRCh38
rs757895216 Chr.9: 137050461-137050461 on Build GRCh38
rs767496302 Chr.9: 137050507-137050507 on Build GRCh38
rs572638976 Chr.9: 137050396-137050396 on Build GRCh38
rs979087022 Chr.9: 137050398-137050398 on Build GRCh38
rs867240992 Chr.9: 137050406-137050406 on Build GRCh38
rs776347357 Chr.9: 137050399-137050399 on Build GRCh38
rs535917485 Chr.9: 137050353-137050353 on Build GRCh38
rs555700532 Chr.9: 137050364-137050364 on Build GRCh38
rs368515415 Chr.9: 137050458-137050458 on Build GRCh38
rs771497567 Chr.9: 137050394-137050394 on Build GRCh38
rs558487969 Chr.9: 137050485-137050485 on Build GRCh38
rs767443128 Chr.9: 137050438-137050438 on Build GRCh38
rs778159229 Chr.9: 137050382-137050382 on Build GRCh38
rs763813336 Chr.9: 137050425-137050425 on Build GRCh38
rs781758869 Chr.9: 137050463-137050463 on Build GRCh38
rs769533874 Chr.9: 137050404-137050404 on Build GRCh38
rs967652291 Chr.9: 137050390-137050391 on Build GRCh38
rs917020422 Chr.9: 137050373-137050373 on Build GRCh38
rs778992568 Chr.9: 137050380-137050380 on Build GRCh38
rs139202937 Chr.9: 137050484-137050484 on Build GRCh38
rs146204639 Chr.9: 137050452-137050452 on Build GRCh38
rs778013442 Chr.9: 137050456-137050456 on Build GRCh38
rs773102878 Chr.9: 137050508-137050508 on Build GRCh38
rs535342828 Chr.9: 137050447-137050447 on Build GRCh38
rs1050344006 Chr.9: 137050412-137050412 on Build GRCh38
rs267602176 Chr.9: 137050459-137050459 on Build GRCh38
rs143867433 Chr.9: 137050532-137050532 on Build GRCh38
rs769215368 Chr.9: 137050473-137050473 on Build GRCh38
rs755262646 Chr.9: 137050366-137050366 on Build GRCh38
rs747541909 Chr.9: 137050390-137050390 on Build GRCh38
rs143961573 Chr.9: 137050514-137050514 on Build GRCh38

View Full Product Details