Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3754021-3754290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
UniGene
Hs.348700
Gene Chromosome Location
Chr.1: 3752398-3771645 on Build GRCh38
Transcripts
NM_152492.2 XM_011540780.1 XM_017000402.1 XM_017000403.1 XM_017000404.1 XM_017000405.1 XM_017000406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805072 c.325C>A p.P109T Chr.1: 3754124-3754124 on Build GRCh38
5380835 c.350G>A p.G117D Chr.1: 3754149-3754149 on Build GRCh38
1667093 c.420G>T p.R140S Chr.1: 3754219-3754219 on Build GRCh38
5380836 c.428C>T p.S143F Chr.1: 3754227-3754227 on Build GRCh38
1686908 c.359C>T p.S120F Chr.1: 3754158-3754158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762493978 Chr.1: 3754191-3754191 on Build GRCh38
rs1020718710 Chr.1: 3754124-3754124 on Build GRCh38
rs376324869 Chr.1: 3754144-3754144 on Build GRCh38
rs188540862 Chr.1: 3754253-3754253 on Build GRCh38
rs770118510 Chr.1: 3754264-3754264 on Build GRCh38
rs772309200 Chr.1: 3754246-3754246 on Build GRCh38
rs1049025361 Chr.1: 3754061-3754061 on Build GRCh38
rs150091953 Chr.1: 3754240-3754240 on Build GRCh38
rs370107628 Chr.1: 3754173-3754173 on Build GRCh38
rs117895136 Chr.1: 3754068-3754068 on Build GRCh38
rs745772584 Chr.1: 3754074-3754074 on Build GRCh38
rs773480563 Chr.1: 3754104-3754104 on Build GRCh38
rs780181258 Chr.1: 3754238-3754238 on Build GRCh38
rs759878876 Chr.1: 3754180-3754180 on Build GRCh38
rs757029536 Chr.1: 3754135-3754135 on Build GRCh38
rs1000546859 Chr.1: 3754134-3754134 on Build GRCh38
rs1012267198 Chr.1: 3754183-3754183 on Build GRCh38
rs777841365 Chr.1: 3754249-3754249 on Build GRCh38
rs149705892 Chr.1: 3754089-3754092 on Build GRCh38
rs765796565 Chr.1: 3754188-3754188 on Build GRCh38
rs773328925 Chr.1: 3754154-3754154 on Build GRCh38
rs775557070 Chr.1: 3754145-3754145 on Build GRCh38
rs776467371 Chr.1: 3754207-3754207 on Build GRCh38
rs771718693 Chr.1: 3754133-3754133 on Build GRCh38
rs757896768 Chr.1: 3754121-3754121 on Build GRCh38
rs141660969 Chr.1: 3754212-3754212 on Build GRCh38
rs776994072 Chr.1: 3754177-3754177 on Build GRCh38
rs200041663 Chr.1: 3754109-3754109 on Build GRCh38
rs879147716 Chr.1: 3754047-3754047 on Build GRCh38
rs754332148 Chr.1: 3754108-3754108 on Build GRCh38
rs145652737 Chr.1: 3754260-3754261 on Build GRCh38
rs866236750 Chr.1: 3754159-3754159 on Build GRCh38
rs768891098 Chr.1: 3754076-3754076 on Build GRCh38
rs1049113436 Chr.1: 3754266-3754266 on Build GRCh38
rs904379094 Chr.1: 3754233-3754233 on Build GRCh38
rs560903163 Chr.1: 3754136-3754136 on Build GRCh38
rs201785236 Chr.1: 3754137-3754137 on Build GRCh38
rs143217722 Chr.1: 3754147-3754147 on Build GRCh38
rs75854782 Chr.1: 3754127-3754127 on Build GRCh38
rs761830156 Chr.1: 3754094-3754094 on Build GRCh38
rs573054510 Chr.1: 3754098-3754098 on Build GRCh38
rs776710502 Chr.1: 3754256-3754256 on Build GRCh38
rs755969582 Chr.1: 3754231-3754231 on Build GRCh38
rs770724512 Chr.1: 3754072-3754072 on Build GRCh38
rs764737784 Chr.1: 3754117-3754117 on Build GRCh38
rs538837971 Chr.1: 3754099-3754099 on Build GRCh38
rs753167186 Chr.1: 3754189-3754189 on Build GRCh38
rs1775812 Chr.1: 3754233-3754233 on Build GRCh38
rs532490794 Chr.1: 3754229-3754229 on Build GRCh38
rs150888098 Chr.1: 3754148-3754148 on Build GRCh38
rs368281276 Chr.1: 3754094-3754097 on Build GRCh38
rs771241421 Chr.1: 3754254-3754254 on Build GRCh38
rs113722627 Chr.1: 3754107-3754107 on Build GRCh38
rs776485155 Chr.1: 3754073-3754073 on Build GRCh38
rs752259546 Chr.1: 3754120-3754120 on Build GRCh38
rs774508129 Chr.1: 3754081-3754081 on Build GRCh38
rs756869473 Chr.1: 3754220-3754220 on Build GRCh38
rs759930374 Chr.1: 3754263-3754263 on Build GRCh38
rs760910706 Chr.1: 3754172-3754172 on Build GRCh38
rs1007910654 Chr.1: 3754086-3754086 on Build GRCh38

View Full Product Details