Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54542872-54543145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL3L
Entrez Gene ID
54552
Gene Name
G protein nucleolar 3 like
Gene Aliases
GNL3B
UniGene
Hs.654677
Gene Chromosome Location
Chr.X: 54530211-54567287 on Build GRCh38
Transcripts
NM_001184819.1 NM_019067.5

SNP Detail

SNP ID Chromosome Location
rs760752301 Chr.X: 54543102-54543102 on Build GRCh38
rs750605652 Chr.X: 54542920-54542920 on Build GRCh38
rs766419351 Chr.X: 54543088-54543088 on Build GRCh38
rs200169500 Chr.X: 54543044-54543044 on Build GRCh38
rs765637571 Chr.X: 54543069-54543069 on Build GRCh38
rs776960227 Chr.X: 54543058-54543058 on Build GRCh38
rs188507576 Chr.X: 54542921-54542921 on Build GRCh38
rs767645257 Chr.X: 54542937-54542937 on Build GRCh38
rs773626886 Chr.X: 54543075-54543075 on Build GRCh38
rs750731737 Chr.X: 54542936-54542937 on Build GRCh38
rs779379084 Chr.X: 54543108-54543108 on Build GRCh38
rs376817364 Chr.X: 54543068-54543068 on Build GRCh38
rs145263040 Chr.X: 54542977-54542977 on Build GRCh38
rs760237282 Chr.X: 54542899-54542899 on Build GRCh38
rs139764430 Chr.X: 54542916-54542916 on Build GRCh38
rs367731494 Chr.X: 54542981-54542981 on Build GRCh38
rs763262088 Chr.X: 54543080-54543080 on Build GRCh38
rs777419878 Chr.X: 54543008-54543008 on Build GRCh38
rs759784443 Chr.X: 54542911-54542911 on Build GRCh38
rs373002660 Chr.X: 54542905-54542905 on Build GRCh38
rs756466044 Chr.X: 54543000-54543002 on Build GRCh38
rs900308876 Chr.X: 54542901-54542901 on Build GRCh38
rs147618956 Chr.X: 54543023-54543023 on Build GRCh38
rs766469132 Chr.X: 54542936-54542936 on Build GRCh38
rs1026632444 Chr.X: 54542915-54542915 on Build GRCh38
rs761243047 Chr.X: 54542904-54542904 on Build GRCh38
rs765214560 Chr.X: 54542961-54542961 on Build GRCh38
rs751492903 Chr.X: 54542938-54542938 on Build GRCh38
rs201258843 Chr.X: 54542945-54542945 on Build GRCh38
rs780557921 Chr.X: 54543049-54543049 on Build GRCh38
rs960377711 Chr.X: 54543000-54543000 on Build GRCh38
rs142243685 Chr.X: 54543027-54543027 on Build GRCh38
rs773877457 Chr.X: 54543033-54543033 on Build GRCh38
rs755842270 Chr.X: 54543007-54543007 on Build GRCh38
rs371840838 Chr.X: 54542999-54542999 on Build GRCh38
rs747514983 Chr.X: 54543043-54543043 on Build GRCh38

View Full Product Details