Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55160260-55160499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20B
Entrez Gene ID
166979
Gene Name
cell division cycle 20B
Gene Aliases
G6VTS76519
UniGene
Hs.669184
Gene Chromosome Location
Chr.5: 55112971-55173177 on Build GRCh38
Transcripts
NM_001145734.2 NM_001170402.1 NM_152623.2 XM_011543218.2 NM_001008397.3 NM_001306197.1 NM_001306198.1 NM_001306201.1 NR_131336.1 NR_131337.1 NR_131338.1 NR_131340.1 XM_006714631.2

Gene Symbol
GPX8
Entrez Gene ID
493869
Gene Name
glutathione peroxidase 8 (putative)
Gene Aliases
EPLA847, GPx-8, GSHPx-8, UNQ847
UniGene
Hs.289044
Gene Chromosome Location
Chr.5: 55160118-55167301 on Build GRCh38
Transcripts
NM_001145734.2 NM_001170402.1 NM_152623.2 XM_011543218.2 NM_001008397.3 NM_001306197.1 NM_001306198.1 NM_001306201.1 NR_131336.1 NR_131337.1 NR_131338.1 NR_131340.1 XM_006714631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379497 c.135C>A p.N45K Chr.5: 55160327-55160327 on Build GRCh38
5507279 c.120C>G p.L40L Chr.5: 55160312-55160312 on Build GRCh38
1068794 c.199G>T p.G67C Chr.5: 55160391-55160391 on Build GRCh38
6674741 c.78T>G p.V26V Chr.5: 55160270-55160270 on Build GRCh38
3344010 c.159G>A p.K53K Chr.5: 55160351-55160351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777226102 Chr.5: 55160423-55160423 on Build GRCh38
rs369372041 Chr.5: 55160401-55160401 on Build GRCh38
rs754103375 Chr.5: 55160402-55160402 on Build GRCh38
rs755647760 Chr.5: 55160346-55160347 on Build GRCh38
rs777117585 Chr.5: 55160421-55160423 on Build GRCh38
rs962288096 Chr.5: 55160470-55160470 on Build GRCh38
rs996685724 Chr.5: 55160378-55160378 on Build GRCh38
rs755405005 Chr.5: 55160414-55160414 on Build GRCh38
rs747684704 Chr.5: 55160342-55160346 on Build GRCh38
rs111518577 Chr.5: 55160344-55160344 on Build GRCh38
rs767762834 Chr.5: 55160371-55160371 on Build GRCh38
rs774635719 Chr.5: 55160359-55160359 on Build GRCh38
rs762826823 Chr.5: 55160346-55160346 on Build GRCh38
rs1015089425 Chr.5: 55160428-55160428 on Build GRCh38
rs764686625 Chr.5: 55160392-55160392 on Build GRCh38
rs373102985 Chr.5: 55160302-55160302 on Build GRCh38
rs761741915 Chr.5: 55160369-55160369 on Build GRCh38
rs769543463 Chr.5: 55160311-55160311 on Build GRCh38
rs915747215 Chr.5: 55160368-55160368 on Build GRCh38
rs754686056 Chr.5: 55160321-55160324 on Build GRCh38
rs768719001 Chr.5: 55160352-55160352 on Build GRCh38
rs200586286 Chr.5: 55160342-55160342 on Build GRCh38
rs772452868 Chr.5: 55160443-55160444 on Build GRCh38
rs780937188 Chr.5: 55160328-55160328 on Build GRCh38
rs746436601 Chr.5: 55160440-55160441 on Build GRCh38
rs928544551 Chr.5: 55160330-55160330 on Build GRCh38
rs998556026 Chr.5: 55160413-55160413 on Build GRCh38
rs374910860 Chr.5: 55160397-55160397 on Build GRCh38

View Full Product Details