Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49927184-49927441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRELD2
Entrez Gene ID
79174
Gene Name
cysteine rich with EGF like domains 2
Gene Aliases
-
UniGene
Hs.211282
Gene Chromosome Location
Chr.22: 49918630-49927540 on Build GRCh38
Transcripts
NM_001135101.2 NM_001284317.1 NM_001284318.1 NM_024324.4 NR_104295.1 XM_005261737.2 XM_017028938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3064468 c.1032G>A p.P344P Chr.22: 49927277-49927277 on Build GRCh38
5859621 c.1024G>A p.E342K Chr.22: 49927269-49927269 on Build GRCh38
3555567 c.1195C>A p.R399S Chr.22: 49927293-49927293 on Build GRCh38
4461133 c.1193C>T p.S398F Chr.22: 49927291-49927291 on Build GRCh38
6635405 c.1051G>A p.E351K Chr.22: 49927296-49927296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745657606 Chr.22: 49927258-49927258 on Build GRCh38
rs1608 Chr.22: 49927295-49927295 on Build GRCh38
rs368886220 Chr.22: 49927294-49927294 on Build GRCh38
rs748419953 Chr.22: 49927240-49927240 on Build GRCh38
rs758721295 Chr.22: 49927216-49927216 on Build GRCh38
rs763742780 Chr.22: 49927271-49927271 on Build GRCh38
rs747538193 Chr.22: 49927231-49927231 on Build GRCh38
rs780439006 Chr.22: 49927230-49927230 on Build GRCh38
rs996320661 Chr.22: 49927222-49927222 on Build GRCh38
rs566966554 Chr.22: 49927204-49927204 on Build GRCh38
rs768743795 Chr.22: 49927325-49927325 on Build GRCh38
rs763285432 Chr.22: 49927281-49927281 on Build GRCh38
rs374713466 Chr.22: 49927274-49927274 on Build GRCh38
rs777875561 Chr.22: 49927301-49927301 on Build GRCh38
rs762636430 Chr.22: 49927348-49927348 on Build GRCh38
rs900564190 Chr.22: 49927393-49927393 on Build GRCh38
rs143871729 Chr.22: 49927277-49927277 on Build GRCh38
rs952312997 Chr.22: 49927397-49927397 on Build GRCh38
rs755457060 Chr.22: 49927293-49927293 on Build GRCh38
rs561634817 Chr.22: 49927323-49927323 on Build GRCh38
rs183678533 Chr.22: 49927396-49927396 on Build GRCh38
rs535311779 Chr.22: 49927244-49927244 on Build GRCh38
rs148204821 Chr.22: 49927287-49927287 on Build GRCh38
rs572595597 Chr.22: 49927283-49927283 on Build GRCh38
rs199838437 Chr.22: 49927205-49927205 on Build GRCh38
rs369179050 Chr.22: 49927334-49927334 on Build GRCh38
rs752469528 Chr.22: 49927221-49927222 on Build GRCh38
rs758053290 Chr.22: 49927319-49927320 on Build GRCh38
rs151286715 Chr.22: 49927273-49927273 on Build GRCh38
rs756156702 Chr.22: 49927296-49927296 on Build GRCh38
rs201734892 Chr.22: 49927346-49927346 on Build GRCh38
rs761366364 Chr.22: 49927276-49927276 on Build GRCh38
rs866859702 Chr.22: 49927269-49927269 on Build GRCh38
rs952258155 Chr.22: 49927259-49927259 on Build GRCh38
rs559149746 Chr.22: 49927313-49927313 on Build GRCh38
rs963933470 Chr.22: 49927369-49927369 on Build GRCh38
rs935244301 Chr.22: 49927341-49927341 on Build GRCh38
rs533714065 Chr.22: 49927394-49927394 on Build GRCh38
rs557186643 Chr.22: 49927250-49927250 on Build GRCh38
rs374692314 Chr.22: 49927349-49927349 on Build GRCh38
rs371917550 Chr.22: 49927314-49927314 on Build GRCh38
rs555904127 Chr.22: 49927232-49927232 on Build GRCh38
rs974665781 Chr.22: 49927401-49927401 on Build GRCh38
rs774369583 Chr.22: 49927226-49927226 on Build GRCh38
rs577045083 Chr.22: 49927340-49927340 on Build GRCh38
rs749516636 Chr.22: 49927309-49927309 on Build GRCh38
rs571117214 Chr.22: 49927234-49927234 on Build GRCh38
rs537870815 Chr.22: 49927359-49927359 on Build GRCh38
rs200272727 Chr.22: 49927264-49927264 on Build GRCh38
rs74379580 Chr.22: 49927415-49927415 on Build GRCh38
rs746748896 Chr.22: 49927321-49927321 on Build GRCh38
rs371759828 Chr.22: 49927335-49927335 on Build GRCh38
rs777182740 Chr.22: 49927374-49927374 on Build GRCh38
rs376500619 Chr.22: 49927333-49927333 on Build GRCh38
rs149445398 Chr.22: 49927260-49927260 on Build GRCh38
rs529933990 Chr.22: 49927375-49927375 on Build GRCh38
rs772807728 Chr.22: 49927347-49927347 on Build GRCh38
rs1040478394 Chr.22: 49927392-49927392 on Build GRCh38

View Full Product Details