Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85710175-85710412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf74
Entrez Gene ID
404550
Gene Name
chromosome 16 open reading frame 74
Gene Aliases
-
UniGene
Hs.461655
Gene Chromosome Location
Chr.16: 85707517-85751083 on Build GRCh38
Transcripts
NM_206967.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852854 c.223G>A p.E75K Chr.16: 85710266-85710266 on Build GRCh38
4141937 c.306G>A p.P102P Chr.16: 85710183-85710183 on Build GRCh38
6407363 c.283A>G p.T95A Chr.16: 85710206-85710206 on Build GRCh38
3755160 c.189A>T p.Q63H Chr.16: 85710300-85710300 on Build GRCh38
6351202 c.192G>A p.M64I Chr.16: 85710297-85710297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747980872 Chr.16: 85710350-85710353 on Build GRCh38
rs115139300 Chr.16: 85710213-85710213 on Build GRCh38
rs111385851 Chr.16: 85710271-85710271 on Build GRCh38
rs374680936 Chr.16: 85710214-85710214 on Build GRCh38
rs965389118 Chr.16: 85710339-85710339 on Build GRCh38
rs369228649 Chr.16: 85710357-85710357 on Build GRCh38
rs761137086 Chr.16: 85710338-85710338 on Build GRCh38
rs555442564 Chr.16: 85710249-85710249 on Build GRCh38
rs1028562371 Chr.16: 85710349-85710349 on Build GRCh38
rs748067415 Chr.16: 85710229-85710229 on Build GRCh38
rs754975130 Chr.16: 85710243-85710243 on Build GRCh38
rs1045690096 Chr.16: 85710279-85710279 on Build GRCh38
rs781002103 Chr.16: 85710307-85710307 on Build GRCh38
rs774927447 Chr.16: 85710322-85710322 on Build GRCh38
rs200347042 Chr.16: 85710297-85710297 on Build GRCh38
rs906204885 Chr.16: 85710254-85710254 on Build GRCh38
rs914072415 Chr.16: 85710375-85710375 on Build GRCh38
rs768084861 Chr.16: 85710269-85710269 on Build GRCh38
rs377716191 Chr.16: 85710227-85710227 on Build GRCh38
rs755848520 Chr.16: 85710384-85710384 on Build GRCh38
rs750689222 Chr.16: 85710270-85710270 on Build GRCh38
rs770628496 Chr.16: 85710197-85710197 on Build GRCh38
rs748838192 Chr.16: 85710325-85710325 on Build GRCh38
rs780331075 Chr.16: 85710221-85710221 on Build GRCh38
rs374677968 Chr.16: 85710335-85710335 on Build GRCh38
rs147002684 Chr.16: 85710361-85710361 on Build GRCh38
rs577189860 Chr.16: 85710316-85710316 on Build GRCh38
rs141454251 Chr.16: 85710300-85710300 on Build GRCh38
rs762344436 Chr.16: 85710210-85710210 on Build GRCh38
rs776284296 Chr.16: 85710257-85710257 on Build GRCh38
rs189450880 Chr.16: 85710240-85710240 on Build GRCh38
rs1018608551 Chr.16: 85710371-85710371 on Build GRCh38
rs767773782 Chr.16: 85710212-85710212 on Build GRCh38
rs766570607 Chr.16: 85710273-85710275 on Build GRCh38
rs201787608 Chr.16: 85710199-85710199 on Build GRCh38
rs779183719 Chr.16: 85710228-85710228 on Build GRCh38
rs375249399 Chr.16: 85710282-85710282 on Build GRCh38
rs762297690 Chr.16: 85710261-85710261 on Build GRCh38
rs372383869 Chr.16: 85710259-85710259 on Build GRCh38
rs368142094 Chr.16: 85710258-85710258 on Build GRCh38
rs759075732 Chr.16: 85710200-85710200 on Build GRCh38
rs369824478 Chr.16: 85710296-85710296 on Build GRCh38
rs775052111 Chr.16: 85710260-85710260 on Build GRCh38
rs374494410 Chr.16: 85710234-85710234 on Build GRCh38
rs997048179 Chr.16: 85710343-85710343 on Build GRCh38
rs745511811 Chr.16: 85710312-85710312 on Build GRCh38
rs919055434 Chr.16: 85710262-85710262 on Build GRCh38
rs1032246010 Chr.16: 85710198-85710198 on Build GRCh38
rs761134549 Chr.16: 85710274-85710274 on Build GRCh38
rs752836530 Chr.16: 85710287-85710287 on Build GRCh38
rs112376688 Chr.16: 85710207-85710207 on Build GRCh38
rs767753044 Chr.16: 85710233-85710252 on Build GRCh38
rs755292101 Chr.16: 85710285-85710285 on Build GRCh38
rs765170842 Chr.16: 85710206-85710207 on Build GRCh38
rs766820240 Chr.16: 85710353-85710353 on Build GRCh38
rs769498389 Chr.16: 85710315-85710315 on Build GRCh38
rs199610353 Chr.16: 85710334-85710334 on Build GRCh38
rs144813253 Chr.16: 85710387-85710387 on Build GRCh38
rs752036541 Chr.16: 85710205-85710205 on Build GRCh38
rs758449740 Chr.16: 85710276-85710278 on Build GRCh38
rs751825593 Chr.16: 85710273-85710281 on Build GRCh38
rs764533571 Chr.16: 85710202-85710202 on Build GRCh38
rs887175499 Chr.16: 85710305-85710305 on Build GRCh38
rs769688679 Chr.16: 85710272-85710273 on Build GRCh38
rs748355555 Chr.16: 85710366-85710366 on Build GRCh38
rs758202202 Chr.16: 85710288-85710288 on Build GRCh38
rs911680439 Chr.16: 85710232-85710232 on Build GRCh38

View Full Product Details