Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41443121-41443392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

Gene Symbol
ERICH4
Entrez Gene ID
100170765
Gene Name
glutamate rich 4
Gene Aliases
C19orf69
UniGene
Hs.649452
Gene Chromosome Location
Chr.19: 41442426-41444765 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960123 c.98C>A p.P33H Chr.19: 41443267-41443267 on Build GRCh38
6579312 c.111C>T p.L37L Chr.19: 41443280-41443280 on Build GRCh38
4519446 c.100G>A p.G34S Chr.19: 41443269-41443269 on Build GRCh38
7316509 c.52G>C p.G18R Chr.19: 41443221-41443221 on Build GRCh38
3227146 c.46G>A p.G16R Chr.19: 41443215-41443215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190183661 Chr.19: 41443322-41443322 on Build GRCh38
rs782160792 Chr.19: 41443226-41443227 on Build GRCh38
rs782153479 Chr.19: 41443213-41443213 on Build GRCh38
rs1016587087 Chr.19: 41443229-41443229 on Build GRCh38
rs556142075 Chr.19: 41443313-41443313 on Build GRCh38
rs782013848 Chr.19: 41443204-41443204 on Build GRCh38
rs1028171138 Chr.19: 41443183-41443183 on Build GRCh38
rs781814529 Chr.19: 41443233-41443234 on Build GRCh38
rs1052938989 Chr.19: 41443155-41443155 on Build GRCh38
rs782775657 Chr.19: 41443168-41443168 on Build GRCh38
rs567420399 Chr.19: 41443227-41443227 on Build GRCh38
rs548701067 Chr.19: 41443169-41443169 on Build GRCh38
rs782756940 Chr.19: 41443270-41443270 on Build GRCh38
rs539595434 Chr.19: 41443337-41443337 on Build GRCh38
rs567385141 Chr.19: 41443214-41443214 on Build GRCh38
rs782463306 Chr.19: 41443233-41443233 on Build GRCh38
rs781851650 Chr.19: 41443272-41443272 on Build GRCh38
rs1000057048 Chr.19: 41443154-41443154 on Build GRCh38
rs782539389 Chr.19: 41443300-41443300 on Build GRCh38
rs782487252 Chr.19: 41443176-41443176 on Build GRCh38

View Full Product Details