Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30601649-30601874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G2E3
Entrez Gene ID
55632
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Gene Aliases
KIAA1333, PHF7B
UniGene
Hs.509008
Gene Chromosome Location
Chr.14: 30559123-30620064 on Build GRCh38
Transcripts
NM_001308097.1 NM_017769.4 XM_011536925.2 XM_011536926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554287 c.857G>T p.R286M Chr.14: 30601874-30601874 on Build GRCh38
2034137 c.810C>T p.C270C Chr.14: 30601827-30601827 on Build GRCh38
6880985 c.757T>G p.W253G Chr.14: 30601774-30601774 on Build GRCh38
333942 c.821G>C p.R274P Chr.14: 30601838-30601838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574903723 Chr.14: 30601848-30601848 on Build GRCh38
rs530942405 Chr.14: 30601672-30601672 on Build GRCh38
rs764183811 Chr.14: 30601786-30601786 on Build GRCh38
rs961449295 Chr.14: 30601681-30601681 on Build GRCh38
rs549011892 Chr.14: 30601669-30601669 on Build GRCh38
rs1057448464 Chr.14: 30601824-30601824 on Build GRCh38
rs768590633 Chr.14: 30601822-30601822 on Build GRCh38
rs139829218 Chr.14: 30601787-30601787 on Build GRCh38
rs770361487 Chr.14: 30601767-30601767 on Build GRCh38
rs766510980 Chr.14: 30601837-30601837 on Build GRCh38
rs756511423 Chr.14: 30601738-30601738 on Build GRCh38
rs189366440 Chr.14: 30601748-30601748 on Build GRCh38
rs537808417 Chr.14: 30601833-30601833 on Build GRCh38
rs778231580 Chr.14: 30601760-30601760 on Build GRCh38
rs775042994 Chr.14: 30601809-30601809 on Build GRCh38
rs778498074 Chr.14: 30601780-30601780 on Build GRCh38
rs936647950 Chr.14: 30601706-30601706 on Build GRCh38
rs912283728 Chr.14: 30601673-30601673 on Build GRCh38
rs758318881 Chr.14: 30601725-30601725 on Build GRCh38
rs945418275 Chr.14: 30601671-30601671 on Build GRCh38
rs753103161 Chr.14: 30601734-30601734 on Build GRCh38
rs972858058 Chr.14: 30601692-30601692 on Build GRCh38
rs374851129 Chr.14: 30601838-30601838 on Build GRCh38
rs931558341 Chr.14: 30601750-30601750 on Build GRCh38
rs920133753 Chr.14: 30601719-30601719 on Build GRCh38
rs371836695 Chr.14: 30601766-30601766 on Build GRCh38
rs776674525 Chr.14: 30601830-30601830 on Build GRCh38
rs760492648 Chr.14: 30601818-30601818 on Build GRCh38

View Full Product Details