Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15782426-15782699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 XM_005274442.4 XM_017029247.1 XM_017029248.1 XM_017029249.1

SNP Detail

SNP ID Chromosome Location
rs369714844 Chr.X: 15782578-15782578 on Build GRCh38
rs763902621 Chr.X: 15782632-15782632 on Build GRCh38
rs753738621 Chr.X: 15782559-15782559 on Build GRCh38
rs749455145 Chr.X: 15782592-15782592 on Build GRCh38
rs950226280 Chr.X: 15782613-15782613 on Build GRCh38
rs769895567 Chr.X: 15782497-15782497 on Build GRCh38
rs755516100 Chr.X: 15782472-15782477 on Build GRCh38
rs199945005 Chr.X: 15782588-15782588 on Build GRCh38
rs763384446 Chr.X: 15782517-15782517 on Build GRCh38
rs371652730 Chr.X: 15782450-15782450 on Build GRCh38
rs757137808 Chr.X: 15782565-15782565 on Build GRCh38
rs12116196 Chr.X: 15782475-15782475 on Build GRCh38
rs372436421 Chr.X: 15782566-15782566 on Build GRCh38
rs747859690 Chr.X: 15782623-15782623 on Build GRCh38
rs774579008 Chr.X: 15782522-15782522 on Build GRCh38
rs774965649 Chr.X: 15782473-15782473 on Build GRCh38
rs768682381 Chr.X: 15782564-15782564 on Build GRCh38
rs1808 Chr.X: 15782628-15782628 on Build GRCh38
rs760211045 Chr.X: 15782557-15782557 on Build GRCh38
rs745741889 Chr.X: 15782466-15782466 on Build GRCh38
rs772685438 Chr.X: 15782639-15782639 on Build GRCh38
rs150953730 Chr.X: 15782529-15782529 on Build GRCh38
rs758252055 Chr.X: 15782571-15782571 on Build GRCh38
rs771333148 Chr.X: 15782595-15782595 on Build GRCh38
rs765052356 Chr.X: 15782640-15782640 on Build GRCh38
rs760825650 Chr.X: 15782659-15782659 on Build GRCh38
rs187583957 Chr.X: 15782558-15782558 on Build GRCh38
rs767259868 Chr.X: 15782549-15782549 on Build GRCh38
rs748193125 Chr.X: 15782493-15782493 on Build GRCh38
rs759792043 Chr.X: 15782600-15782600 on Build GRCh38
rs779838885 Chr.X: 15782663-15782663 on Build GRCh38
rs946610712 Chr.X: 15782556-15782556 on Build GRCh38
rs750161253 Chr.X: 15782569-15782569 on Build GRCh38
rs778966402 Chr.X: 15782567-15782567 on Build GRCh38
rs748393417 Chr.X: 15782581-15782581 on Build GRCh38
rs779199386 Chr.X: 15782657-15782657 on Build GRCh38
rs774974788 Chr.X: 15782596-15782596 on Build GRCh38
rs201165361 Chr.X: 15782555-15782555 on Build GRCh38
rs773661415 Chr.X: 15782498-15782498 on Build GRCh38
rs191657961 Chr.X: 15782671-15782671 on Build GRCh38
rs771261811 Chr.X: 15782520-15782520 on Build GRCh38
rs778573328 Chr.X: 15782462-15782462 on Build GRCh38
rs4830548 Chr.X: 15782477-15782477 on Build GRCh38
rs758142014 Chr.X: 15782645-15782645 on Build GRCh38
rs376671540 Chr.X: 15782625-15782625 on Build GRCh38
rs750404208 Chr.X: 15782641-15782641 on Build GRCh38
rs753274218 Chr.X: 15782638-15782638 on Build GRCh38

View Full Product Details