Gene Transcripts

Gene Symbol
Rom1
Entrez Gene ID
19881
Gene Name
rod outer segment membrane protein 1
Gene Aliases
M101156, Rgsc1156, Rom-1, Rosp1, Tspan23
Chromosome Location
Chr. 19: 8927382 - 8929356 on Build GRCm38
UniGene ID
Mm.426094
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009073.4 NP_033099.3 2 799
GenBank mRNA AK133529.1 - 2 800
BC014827.1 - 2 621 4527997

View Full Product Details