Gene Transcripts

Gene Symbol
Akr1d1
Entrez Gene ID
208665
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
Chromosome Location
Chr. 6: 37529978 - 37568815 on Build GRCm38
UniGene ID
Mm.262635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145364.2 NP_663339.1 8 955
GenBank mRNA BC018333.1 - 8 955

View Full Product Details