Gene Transcripts

Gene Symbol
Arhgap39
Entrez Gene ID
223666
Gene Name
Rho GTPase activating protein 39
Gene Aliases
9530053N22, AI843066, D15Wsu169e, Kiaa1688, Porf-2
Chromosome Location
Chr. 15: 76723985 - 76818170 on Build GRCm38
UniGene ID
Mm.322931
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168288.1 NP_001161760.1 5 1690
NM_198420.2 NP_940812.2 5 1690
GenBank mRNA AK035479.1 - 4 1395
AK141623.1 - 5 1651
AK173232.1 - 5 1625
BC060065.1 - 1 262
BC060637.1 - 5 1671 6849400

View Full Product Details