Gene Transcripts

Gene Symbol
Btnl9
Entrez Gene ID
237754
Gene Name
butyrophilin-like 9
Gene Aliases
B430208I01, Btn3, D330012D11Rik
Chromosome Location
Chr. 11: 49168325 - 49187089 on Build GRCm38
UniGene ID
Mm.130787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172793.2 NP_766381.1 5 951
GenBank mRNA AK080918.1 - 5 935
AK084528.1 - 5 952
AY177687.1 - 6 1221
BC025540.1 - 5 929
BC052556.1 - 4 864
BC151155.1 - 5 908 9088043

View Full Product Details