Gene Transcripts

Gene Symbol
Pcdh11x
Entrez Gene ID
245578
Gene Name
protocadherin 11 X-linked
Gene Aliases
PCDHX, PCDHX11, Pcdh11
Chromosome Location
Chr. X: 120290264 - 120910622 on Build GRCm38
UniGene ID
Mm.39691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271809.1 NP_001258738.1 3 2063
NM_001271810.1 NP_001258739.1 3 2063
GenBank mRNA AF383875.1 - 1 482
AJ564976.1 - 3 2130
BC126930.1 - 3 1986 40086368
EU854474.1 - 1 825

View Full Product Details