Gene Transcripts

Gene Symbol
Abcg5
Entrez Gene ID
27409
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 5
Gene Aliases
AW112016, cmp, sterolin-1, trac
Chromosome Location
Chr. 17: 84658234 - 84682923 on Build GRCm38
UniGene ID
Mm.289590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031884.1 NP_114090.1 7 895
GenBank mRNA AF312713.2 - 7 983
AK149569.1 - 7 866
AY195873.1 - 7 983
BC106766.2 - 7 872 40039484

View Full Product Details