Gene Transcripts

Gene Symbol
Capn13
Entrez Gene ID
381122
Gene Name
calpain 13
Gene Aliases
CANP 13, Gm943
Chromosome Location
Chr. 17: 73306456 - 73399306 on Build GRCm38
UniGene ID
Mm.81496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033444.2 NP_001028616.1 6 803
GenBank mRNA AK136569.1 - 6 804
BC147510.1 - 6 720 9056477
BC147512.1 - 6 720 9056488

View Full Product Details