Gene Transcripts

Gene Symbol
Ahnak2
Entrez Gene ID
100041194
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
AI450948, Gm1185, Gm72
Chromosome Location
Chr. 12: 112772906 - 112802660 on Build GRCm38
UniGene ID
Mm.313169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK140995.1 - 1453
BC138468.1 - 1 1067 8861488
BC138469.1 - 1 1067 8861489

View Full Product Details