Gene Transcripts

Gene Symbol
Prdm14
Entrez Gene ID
383491
Gene Name
PR domain containing 14
Gene Aliases
Chromosome Location
Chr. 1: 13113428 - 13127163 on Build GRCm38
UniGene ID
Mm.309380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081209.2 NP_001074678.2 2 650
GenBank mRNA AK134292.1 - 2 651
AK141218.1 - 1 428
KJ463838.1 - 1 617

View Full Product Details