Gene Transcripts

Gene Symbol
Aadacl2
Entrez Gene ID
639634
Gene Name
arylacetamide deacetylase-like 2
Gene Aliases
EG639634
Chromosome Location
Chr. 3: 60006743 - 60025420 on Build GRCm38
UniGene ID
Mm.475611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128091.1 NP_001121563.1 4, 5 842
GenBank mRNA BC147724.1 - 4, 5 842
BC147728.1 - 4, 5 842 9056677

View Full Product Details