Gene Transcripts

Gene Symbol
Atg13
Entrez Gene ID
51897
Gene Name
autophagy related 13
Gene Aliases
1110053A20Rik, D2Ertd391e, Harbi1
Chromosome Location
Chr. 2: 91674612 - 91710598 on Build GRCm38
UniGene ID
Mm.28492
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145528.3 NP_663503.1 12 1341
GenBank mRNA AK122339.1 - 8 3703
AK149672.1 - 12 1300
AK154470.1 - 12 1329
AK163962.1 - 12 1295
AK171206.1 - 12 1327
AK171440.1 - 12 1318
AK181445.1 - 2 77
AK184342.1 - 2 173
BC016669.1 - 12 1325 2648797
BC023673.1 - 12 1291 5343236
BC023712.1 - 12 1314 5346303
BC033419.1 - 12 1292 5359944

View Full Product Details