Gene Transcripts
- Gene Symbol
- Atg13
- Entrez Gene ID
- 51897
- Gene Name
- autophagy related 13
- Gene Aliases
- 1110053A20Rik, D2Ertd391e, Harbi1
- Chromosome Location
- Chr. 2: 91674612 - 91710598 on Build GRCm38
- UniGene ID
- Mm.28492
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145528.3 |
NP_663503.1
|
12, 13 |
1400 |
|
GenBank mRNA |
AK122339.1 |
- |
8, 9 |
3762 |
|
AK149672.1 |
- |
12, 13 |
1359 |
|
AK154470.1 |
- |
12, 13 |
1388 |
|
AK163962.1 |
- |
12, 13 |
1354 |
|
AK171206.1 |
- |
12, 13 |
1386 |
|
AK171440.1 |
- |
12, 13 |
1377 |
|
AK184342.1 |
- |
2, 3 |
232 |
|
BC016669.1 |
- |
12, 13 |
1384 |
2648797 |
BC023673.1 |
- |
12, 13 |
1350 |
5343236 |
BC023712.1 |
- |
12, 13 |
1373 |
5346303 |
BC033419.1 |
- |
12, 13 |
1351 |
5359944 |
View Full Product Details