Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019780.1 | NP_062754.1 | 2 | 116 | |
GenBank mRNA | AB578902.1 | - | 3 | 126 | |
AB578903.1 | - | 3 | 95 | ||
AB578904.1 | - | 2 | 131 | ||
AB578905.1 | - | 2 | 120 | ||
AB578906.1 | - | 2 | 114 | ||
AB578907.1 | - | 2 | 113 | ||
AB578908.1 | - | 2 | 85 | ||
AB578909.1 | - | 2 | 130 | ||
AB578910.1 | - | 2 | 99 | ||
AB578911.1 | - | 2 | 119 | ||
AF193794.1 | - | 2 | 116 | ||
AK004103.1 | - | 3 | 133 | ||
AK008248.1 | - | 3 | 136 | ||
AK036426.1 | - | 2 | 104 | ||
AK044468.1 | - | 1 | 2438 | ||
AK150330.1 | - | 2 | 126 | ||
BC005663.1 | - | 2 | 105 | 2645432 |