Gene Transcripts

Gene Symbol
Myg1
Entrez Gene ID
60315
Gene Name
melanocyte proliferating gene 1
Gene Aliases
0610023A07Rik, 2810433J21Rik, 5830429P19Rik, AI325965, Gamm1
Chromosome Location
Chr. 15: 102331709 - 102338139 on Build GRCm38
UniGene ID
Mm.251638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021713.2 NP_068359.1 6 958
GenBank mRNA AF289484.1 - 6 950
AK002656.1 - 6 933
AK004104.1 - 6 933
AK013232.1 - 6 904
BC023124.1 - 5 1085
BC028501.1 - 6 943 5256915

View Full Product Details