Gene Transcripts

Gene Symbol
Myg1
Entrez Gene ID
60315
Gene Name
melanocyte proliferating gene 1
Gene Aliases
0610023A07Rik, 2810433J21Rik, 5830429P19Rik, AI325965, Gamm1
Chromosome Location
Chr. 15: 102331709 - 102338139 on Build GRCm38
UniGene ID
Mm.251638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021713.2 NP_068359.1 5, 6 790
GenBank mRNA AF289484.1 - 5, 6 782
AK002656.1 - 5, 6 765
AK004104.1 - 5, 6 765
AK013232.1 - 5, 6 736
BC023124.1 - 4, 5 917
BC028501.1 - 5, 6 775 5256915

View Full Product Details