Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039194.3 | NP_001034283.1 | 5, 6 | 625 | |
NM_001284300.1 | NP_001271229.1 | 5, 6 | 625 | ||
NM_153779.2 | NP_722474.2 | 5, 6 | 618 | ||
NM_178058.4 | NP_835159.1 | 5, 6 | 625 | ||
GenBank mRNA | AK017403.1 | - | 5, 6 | 623 | |
AK085656.1 | - | 5, 6 | 577 | ||
AK147741.1 | - | 5, 6 | 602 | ||
AK155240.1 | - | 5, 6 | 565 | ||
BC038129.1 | - | 5, 6 | 618 | 1395873 |