Gene Transcripts

Gene Symbol
Slc9a8
Entrez Gene ID
77031
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 8
Gene Aliases
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
Chromosome Location
Chr. 2: 167420056 - 167477000 on Build GRCm38
UniGene ID
Mm.278924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB089793.1 - 9 859
AF482993.1 - 8 823
AK049410.1 - 9 855
AK051791.1 - 8 774
AK143281.1 - 9 828
AK160826.1 - 7 697
AK173069.1 - 9 1195
BC030879.1 - 9 858 4241988
BC034508.1 - 2 259
BC058947.1 - 9 829 5702781

View Full Product Details