Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
11593
Gene Name
aspartylglucosaminidase
Gene Aliases
AW060726
Chromosome Location
Chr. 8: 53511687 - 53523422 on Build GRCm38
UniGene ID
Mm.334535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005847.2 NP_001005847.1 2 314
NM_001205054.1 NP_001191983.1 2 314
GenBank mRNA AA396103.1 - 2 159
BC048230.1 - 2 1195
BC132227.1 - 2 289 40130504
BC132229.1 - 2 289 40130506
BC145297.1 - 2 289 9053851
BQ571087.1 - 2 206
CD541877.1 - 2 290
S81393.1 - 2 231

View Full Product Details