Gene Transcripts

Gene Symbol
Aqp5
Entrez Gene ID
11830
Gene Name
aquaporin 5
Gene Aliases
Chromosome Location
Chr. 15: 99591028 - 99594829 on Build GRCm38
UniGene ID
Mm.45580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009701.4 NP_033831.1 4 906
GenBank mRNA AF087654.1 - 4 1155
AK004736.1 - 4 783
AK009302.1 - 4 791
BC026379.1 - 3 1100
BC150769.1 - 5 952 9087680

View Full Product Details