Gene Transcripts

Gene Symbol
Adam25
Entrez Gene ID
23793
Gene Name
a disintegrin and metallopeptidase domain 25 (testase 2)
Gene Aliases
Chromosome Location
Chr. 8: 40752208 - 40756176 on Build GRCm38
UniGene ID
Mm.483208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011781.2 NP_035911.2 2 1297
GenBank mRNA AF167403.1 - 1297
AK016581.1 - 2 1295

View Full Product Details