Gene Transcripts

Gene Symbol
Btla
Entrez Gene ID
208154
Gene Name
B and T lymphocyte associated
Gene Aliases
A630002H24
Chromosome Location
Chr. 16: 45223547 - 45252895 on Build GRCm38
UniGene ID
Mm.38199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY293285.1 - 2 270
BC108963.1 - 2 288 40046744
BC108964.1 - 2 288 40046745

View Full Product Details