Gene Transcripts

Gene Symbol
Nfxl1
Entrez Gene ID
100978
Gene Name
nuclear transcription factor, X-box binding-like 1
Gene Aliases
1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
Chromosome Location
Chr. 5: 72513301 - 72559707 on Build GRCm38
UniGene ID
Mm.187453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133921.2 NP_598682.2 20 2456
GenBank mRNA AK132320.1 - 19 2366
BC022652.1 - 4 456

View Full Product Details