Gene Transcripts

Gene Symbol
Als2cr12
Entrez Gene ID
108812
Gene Name
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12 (human)
Gene Aliases
4933405P16Rik, 4933425F06Rik
Chromosome Location
Chr. 1: 58658117 - 58696010 on Build GRCm38
UniGene ID
Mm.442063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175370.4 NP_780579.1 4, 5 346
GenBank mRNA AK016678.1 - 4, 5 504
AK077190.1 - 4, 5 314
AK133373.1 - 345
BC145039.1 - 3, 4 255

View Full Product Details