Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163505.1 | NP_001156977.1 | 9 | 1153 | |
NM_146091.4 | NP_666203.3 | 9 | 1055 | ||
GenBank mRNA | AK028842.1 | - | 9 | 916 | |
AK030660.1 | - | 9 | 915 | ||
AK077752.1 | - | 9 | 918 | ||
AK147720.1 | - | 9 | 1013 | ||
AK169476.1 | - | 9 | 915 | ||
AK170550.1 | - | 10 | 1006 | ||
BC002149.1 | - | 6 | 445 | ||
BC017138.1 | - | 9 | 904 | 4485115 |