Gene Transcripts

Gene Symbol
Onecut1
Entrez Gene ID
15379
Gene Name
one cut domain, family member 1
Gene Aliases
D9Ertd423e, HNF-6, Hfh12, Hnf6, Hnf6b, OC-1, Oc1
Chromosome Location
Chr. 9: 74861921 - 74889648 on Build GRCm38
UniGene ID
Mm.303355
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008262.3 NP_032288.1 1 1132
GenBank mRNA BC024053.1 - 1 1132
U95945.1 - 1 925

View Full Product Details