Gene Transcripts

Gene Symbol
Olfr33
Entrez Gene ID
18332
Gene Name
olfactory receptor 33
Gene Aliases
MOR11-2, MTPCR33, ORL181
Chromosome Location
Chr. 7: 102713455 - 102714411 on Build GRCm38
UniGene ID
Mm.222837
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147073.1 NP_667284.1 1 306
GenBank mRNA BC104110.1 - 1 306
BC104111.1 - 1 306 40043837

View Full Product Details