Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177647.2 | NP_001171118.1 | 2 | 602 | |
NM_001291019.1 | NP_001277948.1 | 2 | 602 | ||
NM_001291020.1 | NP_001277949.1 | 3 | 404 | ||
NM_001291021.1 | NP_001277950.1 | 2 | 602 | ||
NM_007547.4 | NP_031573.2 | 2 | 602 | ||
GenBank mRNA | AB018194.1 | - | 2 | 168 | |
AF072543.1 | - | 2 | 158 | ||
AF072544.1 | - | 2 | 158 | ||
AF332079.1 | - | 2 | 168 | ||
AF332080.1 | - | 2 | 168 | ||
AK159617.1 | - | 3 | 405 | ||
AK183198.1 | - | 2 | 96 | ||
BC062197.1 | - | 2 | 193 | 5368250 | |
BQ770331.1 | - | 2 | 576 | ||
CD578948.1 | - | 2 | 602 | ||
CX236057.1 | - | 2 | 187 | ||
D87967.1 | - | 2 | 567 | ||
D87968.1 | - | 2 | 567 | ||
U89694.1 | - | 2 | 179 | ||
Y10349.1 | - | 2 | 213 |