Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011327.4 | NP_035457.1 | 14 | 1553 | |
GenBank mRNA | AK002425.1 | - | 3 | 272 | |
AK004860.2 | - | 14 | 1483 | ||
BC018384.1 | - | 14 | 1508 | 3710340 | |
BC034613.1 | - | 14 | 1474 | 5123611 | |
M62361.1 | - | 4 | 458 | ||
M91458.1 | - | 14 | 1468 |