Gene Transcripts

Gene Symbol
Btbd2
Entrez Gene ID
208198
Gene Name
BTB (POZ) domain containing 2
Gene Aliases
2610037C03Rik, 4930512K17Rik
Chromosome Location
Chr. 10: 80642617 - 80657118 on Build GRCm38
UniGene ID
Mm.60720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145361.2 NP_663336.2 2 1018
GenBank mRNA BC055704.1 - 1 85
BC115682.1 - 1 78 40104217
BC115683.1 - 1 78 40104219

View Full Product Details