Gene Transcripts

Gene Symbol
Atxn7l3
Entrez Gene ID
217218
Gene Name
ataxin 7-like 3
Gene Aliases
E030022H21Rik
Chromosome Location
Chr. 11: 102289300 - 102296643 on Build GRCm38
UniGene ID
Mm.390381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098836.1 NP_001092306.1 4, 5 649
NM_001098837.1 NP_001092307.1 4, 5 649
GenBank mRNA AK090368.1 - 4, 5 651
BC130269.1 - 3, 4 399
BM950535.1 - 2, 3 206

View Full Product Details