Gene Transcripts

Gene Symbol
Prdm6
Entrez Gene ID
225518
Gene Name
PR domain containing 6
Gene Aliases
Gm92, PRISM
Chromosome Location
Chr. 18: 53463583 - 53575857 on Build GRCm38
UniGene ID
Mm.297645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033281.3 NP_001028453.1 1 576
GenBank mRNA AK133910.1 - 1 577
BC139295.1 - 1 606 8862317
DQ340565.1 - 1 71

View Full Product Details