Gene Transcripts

Gene Symbol
Lrrc29
Entrez Gene ID
234684
Gene Name
leucine rich repeat containing 29
Gene Aliases
A630024J02Rik, FBL9, FBXL9
Chromosome Location
Chr. 8: 105312340 - 105326331 on Build GRCm38
UniGene ID
Mm.381717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177449.3 NP_803232.2 4 1304
GenBank mRNA AK041606.1 - 5 1807
AK042435.1 - 5 1808
AK148120.1 - 4 1306
BF017011.1 - 1, 2 623

View Full Product Details